Få mere at vide
Mepivacaine-d3, TRC
Artikelnummer. 30473921
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30473921 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473921 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2250572.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Mepivacaine-d3
CAS 1346597-90-7
Molekylær formel C15 D3 H19 N2 O
Mængde 2.5 mg
Synonym N-(2,6-Dimethylphenyl)-1-(trideuteriomethyl)piperidine-2-carboxamide, N-(2,6-Dimethylphenyl)-1-(methyl-d3)-2-piperidinecarboxamide, 1-(Methyl-d3)-2',6'-pipecoloxylidide, (±)-Mepivacaine-d3, Mepivacaine-d3, Mepivacaine-D3 (N-methyl-D3), (±)-Mepivacaine-d3 (N-methyl-d3)
InChI formel InChI=1S/C15H22N2O/c1-11-7-6-8-12(2)14(11)16-15(18)13-9-4-5-10-17(13)3/h6-8,13H,4-5,9-10H2,1-3H3,(H,16,18)/i3D3
SMIL [2H]C([2H])([2H])N1CCCCC1C(=O)Nc2c(C)cccc2C
IUPAC navn N-(2,6-dimethylphenyl)-1-(trideuteriomethyl)piperidine-2-carboxamide
Molekylvægt (g/mol) 249.367
Formel vægt 249.192