Få mere at vide
Mepiperphenidol Bromide, LoGiCal
Artikelnummer. 30462624
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30462624 5 mg
1 options
Artikelnummer. 30462624 Leverandør LoGiCal Leverandørnr. LGCFOR3134.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Mepiperphenidol Bromide
CAS 520-20-7
Analyt- eller komponentnavne Mepiperphenidol Bromide
Forsendelsestilstand Room Temperature
Molekylær formel C19 H32 N O . Br
Certificeringer/compliance ISO 9001
Synonym Piperidinium, 1-(3-hydroxy-5-methyl-4-phenylhexyl)-1-methyl-, bromide (1:1), 1-(3-Hydroxy-5-methyl-4-phenylhexyl)-1-methylpiperidinium bromide (6CI,7CI), Piperidinium, 1-(3-hydroxy-5-methyl-4-phenylhexyl)-1-methyl-, bromide (8CI,9CI), Darstine, Mepiperphenidol, Mepiperphenidol bromide, Mepiperphenidyl, NSC 17126, Piperphenamine
InChI formel InChI=1S/C19H32NO.BrH/c1-16(2)19(17-10-6-4-7-11-17)18(21)12-15-20(3)13-8-5-9-14-20;/h4,6-7,10-11,16,18-19,21H,5,8-9,12-15H2,1-3H3;1H/q+1;/p-1
Løsningstype Neat
SMIL [Br-].CC(C)C(C(O)CC[N+]1(C)CCCCC1)c2ccccc2
IUPAC navn 5-methyl-1-(1-methylpiperidin-1-ium-1-yl)-4-phenylhexan-3-ol;bromide
Industry Type Clinical Testing
Molekylvægt (g/mol) 370.37
Mængde 5 mg
Formel vægt 369.1667 g/mol