Få mere at vide
Mepenzolate Bromide, Mikromol™
Beskrivelse
Mepenzolate Bromide
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Mepenzolate Bromide |
| CAS | 76-90-4 |
| Analyt- eller komponentnavne | Mepenzolate Bromide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C21 H26 N O3 . Br |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Piperidinium, 3-[(2-hydroxy-2,2-diphenylacetyl)oxy]-1,1-dimethyl-, bromide (1:1), 3-Hydroxy-1,1-dimethylpiperidinium bromide benzilate (6CI,7CI), Piperidinium, 3-[(hydroxydiphenylacetyl)oxy]-1,1-dimethyl-, bromide (9CI), Piperidinium, 3-hydroxy-1,1-dimethyl-, bromide, benzilate (8CI), Benzilic acid, ester with 3-hydroxy-1,1-dimethylpiperidinium bromide (8CI), 1-Methyl-3-piperidyl benzilate methylbromide, Cantil, Cantilaque, Cantilon, Cantril, Colibantil, Colopiril, Colum, Delevil, Gastropidil, JB 340, Mepenzolate bromide, N-Methyl-3-piperidyl benzilate methyl bromide, N-Methyl-3-piperidyldiphenylglycolate methobromide, NSC 4358, Trancolon |
| InChI formel | InChI=1S/C21H26NO3.BrH/c1-22(2)15-9-14-19(16-22)25-20(23)21(24,17-10-5-3-6-11-17)18-12-7-4-8-13-18;/h3-8,10-13,19,24H,9,14-16H2,1-2H3;1H/q+1;/p-1 |
| SMIL | [Br-].C[N+]1(C)CCCC(C1)OC(=O)C(O)(c2ccccc2)c3ccccc3 |
| IUPAC navn | (1,1-dimethylpiperidin-1-ium-3-yl) 2-hydroxy-2,2-diphenylacetate;bromide |
| Vis mere |