Få mere at vide
Mepacrine Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Mepacrine Hydrochloride |
| CAS | 69-05-6 |
| Analyt- eller komponentnavne | Mepacrine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C23 H30 Cl N3 O . 2 Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Mepacrine Hydrochloride, 1,4-Pentanediamine, N4-(6-chloro-2-methoxy-9-acridinyl)-N1,N1-diethyl-, hydrochloride (1:2), 1,4-Pentanediamine, N4-(6-chloro-2-methoxy-9-acridinyl)-N1,N1-diethyl-, dihydrochloride (9CI), Acridine, 6-chloro-9-[[4-(diethylamino)-1-methylbutyl]amino]-2-methoxy-, dihydrochloride (8CI), 2-Chloro-5-(ω-diethylamino-α-methylbutylamino)-7-methoxyacridine dihydrochloride, 2-Methoxy-6-chloro-9-(4-diethylamino-1-methylbutylamino)acridine dihydrochloride, 3-Chloro-7-methoxy-9-(1-methyl-4-diethylaminobutylamino)acridine dihydrochloride, 3-Chloro-9-(4'-diethylamino-1'-methylbutylamino)-7-methoxyacridine dihydrochloride, 6-Chloro-9-[[4-(diethylamino)-1-methylbutyl]amino]-2-methoxyacridine dihydrochloride, 866RP, Acrichin dihydrochloride, Atabrine hydrochloride, Atebrin, Atebrine, Atebrine hydrochloride, Chemiochin, Chinacrin hydrochloride, Crinodora, Erion, Malaricida, Mecryl, Mepacrine dihydrochloride, Mepacrine hydrochloride, Methoquine, Metochin, Metoquin, Metoquine, NSC 14229, Palacrin, Palusan, Pentilen, Quinacrine dihydrochloride, Quinacrine hydrochloride, SN 390 |
| InChI formel | InChI=1S/C23H30ClN3O.2ClH/c1-5-27(6-2)13-7-8-16(3)25-23-19-11-9-17(24)14-22(19)26-21-12-10-18(28-4)15-20(21)23;;/h9-12,14-16H,5-8,13H2,1-4H3,(H,25,26);2*1H |
| Løsningstype | Neat |
| SMIL | Cl.Cl.CCN(CC)CCCC(C)Nc1c2ccc(Cl)cc2nc3ccc(OC)cc13 |
| Vis mere |