Få mere at vide
Menthol-d4 (mixture of diastereomers), TRC
Artikelnummer. 30470214
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30470214 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470214 Leverandør Toronto Research Chemicals Leverandørnr. TRCM218902100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Menthol-d4 (mixture of diastereomers)
CAS 1217765-02-0
Molekylær formel C10H16D4O
Mængde 100 mg
Synonym 5-Methyl-2-(1-methylethyl)cyclohexanol-d4, 1-Methyl-4-isopropyl-3-cyclohexanol-d4, 2-Isopropyl-5-methylcyclohexan-1-ol-d4, 2-Isopropyl-5-methylcyclohexanol-d4, 3-Hydroxy-p-menthane-d4, 5-Methyl-2-(1-methylethyl)cyclohexanol-d4, 5-Methyl-2-isopropylcyclohexanol-d4, Menthyl Alcohol-d4, p-Menthan-3-ol-d4
InChI formel InChI=1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/i6D2,9D,10D
SMIL [2H]C1(C(C)C)CCC(C)C([2H])([2H])C1(O)[2H]
Molekylvægt (g/mol) 160.29
Formel vægt 160.1765