Få mere at vide
Menaquinone 7-d7(>90%), TRC
Artikelnummer. 30465384
Skift visning
Mængde:
0.25 mg
Artikelnummer. Mængde
30465384 0.25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465384 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2186020.25MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Menaquinone 7-d7
CAS 1233937-31-9
Molekylær formel C46H57D7O2
Mængde 0.25 mg
Synonym (all-E)-2-(3,7,11,15,19,23,27-Heptamethyl-2,6,10,14,18,22,26-octacosaheptaenyl)-3-methyl-1,4-naphthalenedione-d7, Vitamin K2(35)-d7, Menaquinone K7-d7, Vitamin MK 7-d7
InChI formel InChI=1S/C46H64O2/c1-34(2)18-12-19-35(3)20-13-21-36(4)22-14-23-37(5)24-15-25-38(6)26-16-27-39(7)28-17-29-40(8)32-33-42-41(9)45(47)43-30-10-11-31-44(43)46(42)48/h10-11,18,20,22,24,26,28,30-32H,12-17,19,21,23,25,27,29,33H2,1-9H3/b35-20+,36-22+,37-24+,38-26+,39-28+,40-32+/i9D3,10D,11D,30D,31D
SMIL O=C1C(C/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/C)=C(C([2H])([2H])[2H])C(C2=C([2H])C([2H])=C([2H])C([2H])=C21)=O
Molekylvægt (g/mol) 656.04
Formel vægt 655.5346