Få mere at vide
Menaquinone 4-d7 (Mixture of cis-trans isomers), TRC
Artikelnummer. 30475738
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30475738 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30475738 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2185972.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Menaquinone 4-d7 (Mixture of cis-trans isomers)
CAS 1233937-25-1
Renhedsgrad noter HPLC
Molekylær formel C31H33D7O2
Mængde 2.5 mg
Synonym (E,E,E)-2-Methyl-3-(3,7,11,15-tetramethyl-2,6,10,14-hexadecatetraenyl)-1,4-naphthalenedione-d7, 2-Methyl-3-geranylgeranyl-1,4-naphthoquinone-d7, Glakay-d7, Kaytwo-d7, MK4-d7, Menaquinone K4-d7, Menatetrenone-d7, Vitamin K2(20)-d7, Vitamin MK 4-d7
InChI formel InChI=1S/C31H40O2/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-17-25(5)20-21-27-26(6)30(32)28-18-7-8-19-29(28)31(27)33/h7-8,12,14,16,18-20H,9-11,13,15,17,21H2,1-6H3/b23-14+,24-16+,25-20+/i6D3,7D,8D,18D,19D
SMIL O=C1C(C/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/C)=C(C([2H])([2H])[2H])C(C2=C([2H])C([2H])=C([2H])C([2H])=C21)=O
Molekylvægt (g/mol) 451.69
Formel vægt 451.3468
Procent renhed >95