Learn More
Memory Test Solution 1 For ICP/MS, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep CL-MEM-1
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Al | 1000ÎĽg/mL |
Ca | 1000ÎĽg/mL |
Fe | 1000ÎĽg/mL |
K | 1000ÎĽg/mL |
Mg | 1000ÎĽg/mL |
Na | 1000ÎĽg/mL |
Ag | 20ÎĽg/mL |
As | 20ÎĽg/mL |
Ba | 20ÎĽg/mL |
Be | 20ÎĽg/mL |
Cd | 20ÎĽg/mL |
Co | 20ÎĽg/mL |
Cr | 20ÎĽg/mL |
Cu | 20ÎĽg/mL |
Mn | 20ÎĽg/mL |
Ni | 20ÎĽg/mL |
Pb | 20ÎĽg/mL |
Se | 20ÎĽg/mL |
Tl | 20ÎĽg/mL |
V | 20ÎĽg/mL |
Zn | 20ÎĽg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Memory Test 1 | |
125 mL | |
ICP-MS | |
Standard Solution | |
5% HNO3 | |
Components in 5% HNO3: Al •1000μg/mL As •20μg/mL Ba •20μg/mL Be •20μg/mL Cd •20μg/mL Ca •1000μg/mL Co •20μg/mL Cr •20μg/mL Cu •20μg/mL Fe •1000μg/mL Pb •20μg/mL Mg •1000μg/mL Mn •20μg/mL Ni •20μg/mL K •1000μg/mL Se •20μg/mL Ag •20μg/mL Na •1000μg/mL Tl •20μg/mL V •20μg/mL Zn •20μg/mL |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
ICP/ICP-MS Standard | |
ICP/ICP-MS |
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitric acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product intended for laboratory use only