Få mere at vide
Memantine Glycine, TRC
Artikelnummer. 30480855
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480855 10 mg
30480663 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30480855 Leverandør Toronto Research Chemicals Leverandørnr. TRCM21801510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1340545-88-1
Molekylær formel C14 H23 N O2
Molekylvægt (g/mol) 237.34
Synonym Glycine, N-(3,5-dimethyltricyclo[3.3.1.13,7]dec-1-yl)-, N-(3,5-Dimethyltricyclo[3.3.1.13,7]dec-1-yl)glycine
IUPAC navn 2-[(3,5-dimethyl-1-adamantyl)amino]acetic acid
SMIL CC12CC3CC(C)(C1)CC(C3)(C2)NCC(=O)O
Tekniske data
Kemisk navn eller materiale Memantine Glycine
InChI formel InChI=1S/C14H23NO2/c1-12-3-10-4-13(2,7-12)9-14(5-10,8-12)15-6-11(16)17/h10,15H,3-9H2,1-2H3,(H,16,17)
Mængde 10 mg
Formel vægt 237.1729