Få mere at vide
Memantine-d6 Hydrochloride, TRC
Artikelnummer. 30482873
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30482873 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482873 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2180021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Memantine-d6 Hydrochloride
CAS 1189713-18-5
Renhedsgrad noter HPLC
Molekylær formel C12 2H6 H15 N . Cl H
Mængde 1 mg
Synonym 3,5-Di(methyl-d3)-tricyclo[3.3.1.13,7]decan-1-amine Hydrochloride (1:1), Memantin-d6-Dimethyl
InChI formel InChI=1S/C12H21N.ClH/c1-10-3-9-4-11(2,6-10)8-12(13,5-9)7-10;/h9H,3-8,13H2,1-2H3;1H/i1D3,2D3;
SMIL Cl.[2H]C([2H])([2H])C12CC3CC(N)(CC(C3)(C1)C([2H])([2H])[2H])C2
IUPAC navn 3,5-bis(trideuteriomethyl)adamantan-1-amine;hydrochloride
Molekylvægt (g/mol) 221.8
Formel vægt 221.1817
Procent renhed >95