Få mere at vide
Gibco™ MEM, Hanks' Balanced Salts, powder
Beskrivelse
Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing
Gibco™ MEM is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). This MEM formulation does not require a CO2 environment to maintain physiological pH.
Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing
Gibco™ MEM is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). This MEM formulation does not require a CO2 environment to maintain physiological pH.
Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).
Tekniske data
Tekniske data
| Form | Powder |
| Produkttype | Balanced Salts MEM |
| Med tilsætningsstoffer | Hanks’ Balanced Salt solution (HBSS), Glutamine, Phenol Red, Low Glucose, HEPES |
| Uden tilsætningsstoffer | No Sodium Bicarbonate |
| Til brug med (applikation) | Mammalian cell culture |
| Mængde | 50 l |
| Certificeringer/compliance | ISO 13485 standards, FDA |
| Regulatorisk status | For research use only |
| Indhold og opbevaring | 2 to 8°C, room temperature |
| Oversigt | Balanced Salts MEM, 50L, Powder, 36 Months Shelf Life, Hanks' Salts, L-Glutamine, Phenol Red and HEPS with Additives, Sodium Bicarbonate Without Additives, Ambient Shipping Condition, 2 to 8°C Storage Requirements, For Mammalian Cell Culture |