Få mere at vide
Melphalan-d8 Monohydrochloride, TRC
Artikelnummer. 30179351
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179351 1 mg
30179370 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30179351 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2169031MG
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1217854-43-7
Molekylær formel C13H10D8Cl2N2O2 . HCl
Molekylvægt (g/mol) 349.71
Synonym 4-[Bis(2-chloroethyl)amino]-L-phenylalanine-d8 Monohydrochloride, p-Di-(2-chloroethyl)amino-L-phenylalanine-d8 Monohydrochloride, L-Phenylalanine Mustard-d8 Monohydrochloride, Melfalan-d8 Monohydrochloride;
IUPAC navn (S)-2-amino-3-(4-(bis(2-chloroethyl-1,1,2,2-d4)amino)phenyl)propanoic acid hydrochloride
SMIL [2H]C(C([2H])(Cl)[2H])(N(C([2H])(C([2H])(Cl)[2H])[2H])C1=CC=C(C[C@H](N)C(O)=O)C=C1)[2H].Cl
Tekniske data
Kemisk navn eller materiale Melphalan-d8 Hydrochloride
Mængde 1 mg
InChI formel InChI=1S/C13H18Cl2N2O2.ClH/c14-5-7-17(8-6-15)11-3-1-10(2-4-11)9-12(16)13(18)19;/h1-4,12H,5-9,16H2,(H,18,19);1H/t12-;/m0./s1/i5D2,6D2,7D2,8D2;
Formel vægt 348.1