Få mere at vide
Melperone, LoGiCal
Artikelnummer. 30459025
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459025 10 mg
1 options
Artikelnummer. 30459025 Leverandør LoGiCal Leverandørnr. LGCFOR0321.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Melperone
CAS 3575-80-2
Analyt- eller komponentnavne Melperone
Forsendelsestilstand Room Temperature
Molekylær formel C16 H22 F N O
Certificeringer/compliance ISO 9001
Synonym 1-Butanone, 1-(4-fluorophenyl)-4-(4-methyl-1-piperidinyl)-, Butyrophenone, 4'-fluoro-4-(4-methylpiperidino)- (7CI,8CI), g-(4-Methylpiperidino)-p-fluorobutyrophenone, 4'-Fluoro-4-(4-methylpiperidino)butyrophenone, 4-Fluoro-g-(4-methylpiperidino)butyrophenone, Buronil, FG 5111, Flubuperone, Melperone, Methylperone, g-(4-Methylpiperidino)-p-fluorobutyrophenone, 4-Fluoro-g-(4-methylpiperidino)butyrophenone
InChI formel InChI=1S/C16H22FNO/c1-13-8-11-18(12-9-13)10-2-3-16(19)14-4-6-15(17)7-5-14/h4-7,13H,2-3,8-12H2,1H3
Løsningstype Neat
SMIL CC1CCN(CCCC(=O)c2ccc(F)cc2)CC1
IUPAC navn 1-(4-fluorophenyl)-4-(4-methylpiperidin-1-yl)butan-1-one
Industry Type Clinical Testing
Molekylvægt (g/mol) 263.35
Mængde 10 mg
Formel vægt 263.1685 g/mol