Få mere at vide
Meloxicam D3 (2-methyl D3) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Artikelnummer. 30324861
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
30324861 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324861 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA14862510AL100
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Meloxicam D3 (2-methyl D3)
CAS 942047-63-4
Opløsningsmiddel eller Matrix Acetonitrile
Molekylær formel C14 2H3 H10 N3 O4 S2
Mængde 1 mL
Synonym Meloxicam D3 (2-methyl D3)
InChI formel InChI=1S/C14H13N3O4S2/c1-8-7-15-14(22-8)16-13(19)11-12(18)9-5-3-4-6-10(9)23(20,21)17(11)2/h3-7,18H,1-2H3,(H,15,16,19)/i2D3
SMIL [2H]C([2H])([2H])N1C(=C(O)c2ccccc2S1(=O)=O)C(=O)Nc3ncc(C)s3
IUPAC navn 4-hydroxy-N-(5-methyl-1,3-thiazol-2-yl)-1,1-dioxo-2-(trideuteriomethyl)-1λ^{6},2-benzothiazine-3-carboxamide
Molekylvægt (g/mol) 354.42
Formel vægt 354.0536 g/mol