Få mere at vide
Meloxicam- 13CD3, TRC
Artikelnummer. 30484212
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30484212 10 mg
30473424 1 mg
30476914 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30484212 Leverandør Toronto Research Chemicals Leverandørnr. TRCM21610310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1309936-00-2
Molekylær formel C1313CH10D3N3O4S2
Molekylvægt (g/mol) 355.41
Synonym 4-Hydroxy-N-(5-methyl-1,3-thiazol-2-yl)-1,1-dioxo-2-(trideuteriomethyl)-1lambda{6},2-benzothiazine-3-carboxamide
IUPAC navn 4-hydroxy-N-(5-methyl-1,3-thiazol-2-yl)-1,1-dioxo-2-(trideuterio(1^{13}C)methyl)-1λ^{6},2-benzothiazine-3-carboxamide
SMIL [2H][13C]([2H])([2H])N1C(=C(O)c2ccccc2S1(=O)=O)C(=O)Nc3ncc(C)s3
Tekniske data
Kemisk navn eller materiale Meloxicam- 13CD3
Mængde 10 mg
InChI formel InChI=1S/C14H13N3O4S2/c1-8-7-15-14(22-8)16-13(19)11-12(18)9-5-3-4-6-10(9)23(20,21)17(11)2/h3-7,18H,1-2H3,(H,15,16,19)/i2+1D3
Formel vægt 355.0569