Få mere at vide
MedChemExpress Melittin
Artikelnummer. 16442699
Encompass
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
16442699 1 mg
1 options
Artikelnummer. 16442699 Leverandør MedChemExpress Leverandørnr. HYP0233
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
MedChemExpress Melittin is a PLA2 activator, stimulates the activity of the low molecular weight PLA2, while it does not the increase activity of the high molecular weight PLA2.
Tekniske data
CAS 20449-79-0
Indhold og opbevaring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Til brug med (applikation) Cancer-programmed cell death
Molekylvægt (g/mol) 2846.46 g/mol
Produkttype Peptides