Få mere at vide
Melitracen-d6 Hydrochloride, TRC
Artikelnummer. 30468235
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30468235 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468235 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2155021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Melitracen-d6 Hydrochloride
CAS 1189648-08-5
Molekylær formel C21H20D6ClN
Mængde 1 mg
Synonym 3-(10,10-Dimethyl-d6)-9(10H)-anthracenylidene)-N,N-dimethyl-1-propanamine Hydrochloride, 9-(3-Dimethylaminopropylidene)-10,10-(dimethyl-d6) -9,10-dihydroanthracene Hydrochloride, Adaptol-d6, Melixeran-d6, Melitracene-d6, Thymeol-d6, Trausabun-d6, U-24973A-d6
InChI formel InChI=1S/C21H25N.ClH/c1-21(2)19-13-7-5-10-17(19)16(12-9-15-22(3)4)18-11-6-8-14-20(18)21;/h5-8,10-14H,9,15H2,1-4H3;1H/i1D3,2D3;
SMIL CN(C)CC/C=C1C2=C(C=CC=C2)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C3=CC=CC=C3\1.Cl
Molekylvægt (g/mol) 333.93