Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Melengestrol-acetate |
| CAS | 2919-66-6 |
| CAS Max % | ≤ 100% |
| Beilstein | 2066919 |
| Farve | White/Yellow |
| Smeltepunkt | 216°C to 223°C |
| Kogepunkt | 424°C |
| Synonym | 17α-Acetoxy-6-methyl-16-methylene-4,6-pregnadiene-3,20-dione, MAG |
| Molekylvægt (g/mol) | 396.52 g/mol |
| Mængde | 100 mg |