missing translation for 'onlineSavingsMsg'
Få mere at vide

Melarsoprol (Mixture of Diastereomers), TRC

Artikelnummer. 30136863
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136863 250 mg
30004622 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Artikelnummer. 30136863 Leverandør Toronto Research Chemicals Leverandørnr. TRCM215100250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 494-79-1
Molekylær formel C12 H15 As N6 O S2
Molekylvægt (g/mol) 398.34
Synonym 1,3,2-Dithiarsolane-4-methanol, 2-[4-[(4,6-diamino-1,3,5-triazin-2-yl)amino]phenyl]-, 1,3,2-Dithiarsolane-4-methanol, 2-[p-[(4,6-diamino-s-triazin-2-yl)amino]phenyl]- (8CI), Benzenearsonous acid, p-[(4,6-diamino-s-triazin-2-yl)amino]dithio-, cyclic (hydroxymethyl)ethylene ester (6CI,7CI), 2-[4-[(4,6-Diamino-1,3,5-triazin-2-yl)amino]phenyl]-1,3,2-dithiarsolane-4-methanol, 2-[p-(4,6-Diamino-s-triazin-2-ylamino)phenyl]-1,3,2-dithiarsolane-4-methanol, 2-p-(4,6-Diamino-s-triazin-2-ylamino)phenyl-4-hydroxymethyl-1,3,2-dithiarsoline, 3854RP, Arsobal, Arsorbal, Benzenearsonous acid, p-[(4,6-diamino-s-triazin-2-yl)amino]dithio-, (hydroxymethyl)ethylene ester, Mel B, Mel B Arsobal, Melarsaprol, Melarsen B, Melarsoprol, Melarsoprol B, RP 3854
IUPAC navn [2-[4-[(4,6-diamino-1,3,5-triazin-2-yl)amino]phenyl]-1,3,2-dithiarsolan-4-yl]methanol
SMIL Nc1nc(N)nc(Nc2ccc(cc2)[As]3SCC(CO)S3)n1

Tekniske data

Kemisk navn eller materiale Melarsoprol (Mixture of Diastereomers)
Mængde 250 mg
InChI formel InChI=1S/C12H15AsN6OS2/c14-10-17-11(15)19-12(18-10)16-8-3-1-7(2-4-8)13-21-6-9(5-20)22-13/h1-4,9,20H,5-6H2,(H5,14,15,16,17,18,19)
Formel vægt 397.9965
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.