Få mere at vide
Melarsen Oxide, TRC
Artikelnummer. 30482207
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482207 50 mg
30476438 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30482207 Leverandør Toronto Research Chemicals Leverandørnr. TRCM21511050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 21840-08-4
Molekylær formel C9H9AsN6O
Molekylvægt (g/mol) 292.13
Synonym N2-(4-Arsenosophenyl)-1,3,5-triazine-2,4,6-triamine, (p-Arsenosophenyl)melamine, (p-Arsenosophenyl)melamine, 2-(p-Arsenosoanilino)-4,6-diamino-s-triazine, 3177 R.P., Friedheim 2065, Melarsenoxyd, Melox, NSC 34642
IUPAC navn 2-N-(4-arsorosophenyl)-1,3,5-triazine-2,4,6-triamine
SMIL Nc1nc(N)nc(Nc2ccc(cc2)[As]=O)n1
Tekniske data
Kemisk navn eller materiale Melarsen Oxide
Mængde 50 mg
InChI formel InChI=1S/C9H9AsN6O/c11-7-14-8(12)16-9(15-7)13-6-3-1-5(10-17)2-4-6/h1-4H,(H5,11,12,13,14,15,16)
Formel vægt 292.0054