missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Melanotransferrin/CD228/MFI2 Antibody (SC-005) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28837JF635
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Melanotransferrin/CD228/MFI2 Monoclonal antibody specifically detects Melanotransferrin/CD228/MFI2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
| Melanotransferrin/CD228/MFI2 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and96.5, CD228, CD228 antigen, FLJ38863, MAP97Melanoma-associated antigen p97, melanotransferrin, membrane-bound transferrin-like protein, MGC4856, MTF1 | |
| Melanotransferrin/ CD228 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| SC-005 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| 4241 | |
| Store at 4°C in the dark. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur