Få mere at vide
3M™ Melamine Handpad
Artikelnummer. 17267630 Shop alle 3M produkter
Skift visning
Længde (metrisk):
115 mm
Artikelnummer. Længde (metrisk)
17267630 115 mm
1 options
Artikelnummer. 17267630 Leverandør 3M™ Leverandørnr. 7100020667
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Melamine Handpad
  • Developed for use on stone, concrete, porcelain stoneware, marble and other porous floors and is used to remove deep-seated dirt and unsightly traces of abrasion
  • Superpad for modern microporous floor surfaces such as ceramic, terracotta, marble and concrete
  • Removes most dirt and stains with just water
  • More environmentally friendly - no chemical additives or cleaning agents required
TRUSTED_SUSTAINABILITY
Tekniske data
Længde (metrisk) 115 mm
Bredde (metrisk) 250 mm