Få mere at vide
MEIS2 Rabbit anti-Human, Mouse, Clone: 2A6I3, R&D Systems™
Artikelnummer. 18310702
Skift visning
Mængde:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18310702 100 μg
18369334 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18310702 Leverandør Bio-Techne Leverandørnr. NBP316224100UL
 Begrænset vare
Forventet forsendelse: 23-10-2026
A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody
MEIS2 Monoclonal specifically detects MEIS2 in Human∣Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen MEIS2
Applikationer Western Blot
Klassifikation Monoclonal
Klon 2A6I3
Konjugeret Unconjugated
Formulering PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Meis (mouse) homolog 2, Meis homeobox 2, Meis1, myeloid ecotropic viral integration site 1 homolog 2 (mouse), Meis1-related gene 1, MGC2820, myeloid ecotropic viral integration site 1 homolog 2, TALE homeobox protein Meis2
Værtsarter Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MEIS2 (O14770).
Oprensningsmetode Affinity purified
Mængde 100 μg
Regulatorisk status RUO
Forskningsdisciplin Vision
Primær eller sekundær Primary
Gen-id (Entrez) 4212
Målarter Human, Mouse
Indhold og opbevaring Store at -20°C. Avoid freeze-thaw cycles.
Produkttype Antibody
Form Purified
Isotype IgG
Produktlinje Novus