Få mere at vide
Megestrol-d3, TRC
Artikelnummer. 30483443
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30483443 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483443 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2080421MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Megestrol-d3
CAS 162462-71-7
Molekylær formel C22H27D3O3
Mængde 1 mg
Synonym 17-Hydroxy-6-(methyl-d3)-pregna-4,6-diene-3,20-dione, 6-(Methyl-d3)megestrol
InChI formel InChI=1S/C22H30O3/c1-13-11-16-17(20(3)8-5-15(24)12-19(13)20)6-9-21(4)18(16)7-10-22(21,25)14(2)23/h11-12,16-18,25H,5-10H2,1-4H3/t16-,17+,18+,20-,21+,22+/m1/s1/i1D3
SMIL [2H]C([2H])([2H])C1=C[C@@H]2[C@H](CC[C@@]3(C)[C@H]2CC[C@]3(O)C(=O)C)[C@@]4(C)CCC(=O)C=C14
IUPAC navn (8R,9S,10R,13S,14S,17R)-17-acetyl-17-hydroxy-10,13-dimethyl-6-(trideuteriomethyl)-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 345.49
Formel vægt 345.2383