Learn More
Mefloquine Hydrochloride, LoGiCal
Product Code. 30464288
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30464288 10 mg 10mg
1 options
Product Code. 30464288 Supplier LoGiCal Supplier No. LGCFOR1198.00
 Restricted Item
Forventet forsendelse: 27-11-2026
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Mefloquine hydrochloride
CAS 51773-92-3
Analyte or Component Names Mefloquine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C17 H16 F6 N2 O . Cl H
Certifications/Compliance ISO 9001
Synonym Mefloquine hydrochloride, (RS)-[2,8-Bis(trifluoromethyl)quinolin-4-yl][(2SR)-piperidin-2-yl]methanol hydrochloride
InChI Formula InChI=1S/C17H16F6N2O.ClH/c18-16(19,20)11-5-3-4-9-10(15(26)12-6-1-2-7-24-12)8-13(17(21,22)23)25-14(9)11;/h3-5,8,12,15,24,26H,1-2,6-7H2;1H/t12-,15+;/m0./s1
Solution Type Neat
SMILES Cl.O[C@@H]([C@@H]1CCCCN1)c2cc(nc3c(cccc23)C(F)(F)F)C(F)(F)F
IUPAC Name (R)-[2,8-bis(trifluoromethyl)quinolin-4-yl]-[(2S)-piperidin-2-yl]methanol;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 414.77
Quantity 10 mg
Formula Weight 414.0934 g/mol