Få mere at vide
Mefloquine EP Impurity C (as Hydrochloride), Mikromol™
Beskrivelse
Mefloquine EP Impurity C (as Hydrochloride), Mikromol
Tekniske data
Tekniske data
| Kemisk navn eller materiale | threo-Mefloquine Hydrochloride |
| CAS | 51744-85-5 |
| Analyt- eller komponentnavne | threo-Mefloquine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17H16F6N2O . HCl |
| Certificeringer/compliance | ISO 17025 |
| Synonym | (RS)-[2,8-Bis(trifluoromethyl)quinolin-4-yl][(2RS)-piperidin-2-yl]methanol Hydrochloride (threo-Mefloquine Hydrochloride), [S-(R*,R*)]-α-2-piperidinyl-2,8-bis(trifluoromethyl)-4-Quinolinemethanol Monohydrochloride, USP Mefloquine Related Compound A; |
| InChI formel | InChI=1S/C17H16F6N2O.ClH/c18-16(19,20)11-5-3-4-9-10(15(26)12-6-1-2-7-24-12)8-13(17(21,22)23)25-14(9)11;/h3-5,8,12,15,24,26H,1-2,6-7H2;1H/t12-,15-;/m0./s1 |
| Løsningstype | Neat |
| SMIL | [H][C@@]1(NCCCC1)[C@@H](O)C2=CC(C(F)(F)F)=NC3=C(C(F)(F)F)C=CC=C32.Cl |
| Vis mere |