Få mere at vide
Mefenpyr-diethyl, TRC
Artikelnummer. 30476946
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30476946 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476946 Leverandør Toronto Research Chemicals Leverandørnr. TRCM205603250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Mefenpyr-diethyl
CAS 135590-91-9
Renhedsgrad noter HPLC
Molekylær formel C16 H18 Cl2 N2 O4
Mængde 250 mg
Synonym 1H-Pyrazole-3,5-dicarboxylic acid, 1-(2,4-dichlorophenyl)-4,5-dihydro-5-methyl-, diethyl ester (9CI), AE-F 107892, Ethyl 1-(2,4-dichlorophenyl)-5-(ethoxycarbonyl)-5-methyl-2-pyrazoline-3-carboxylate, HOE 107892, Mefenpyr-diethyl
InChI formel InChI=1S/C16H18Cl2N2O4/c1-4-23-14(21)12-9-16(3,15(22)24-5-2)20(19-12)13-7-6-10(17)8-11(13)18/h6-8H,4-5,9H2,1-3H3
SMIL CCOC(=O)C1=NN(c2ccc(Cl)cc2Cl)C(C)(C1)C(=O)OCC
IUPAC navn diethyl 1-(2,4-dichlorophenyl)-5-methyl-4H-pyrazole-3,5-dicarboxylate
Molekylvægt (g/mol) 373.23
Formel vægt 372.0644
Procent renhed >95