Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Mefenpyr-diethyl |
| Emballage | Glass Bottle |
| CAS | 135590-91-9 |
| Farve | Beige |
| Smeltepunkt | 50°C to 52°C |
| Kogepunkt | 260°C (Decomposition) |
| Synonym | diethyl 1-(2,4-dichlorophenyl)-4,5-dihydro-5-methyl-1H-pyrazole-3,5-dicarboxylate |
| Industry Type | Environmental Analysis |
| Molekylvægt (g/mol) | 373.23 |
| Mængde | 100 mg |
| Vis mere |