Få mere at vide
Mefenamic Acid EP Impurity B, Mikromol™
Artikelnummer. 30052390
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052390 100 mg
30051698 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052390 Leverandør Mikromol Leverandørnr. MM0327.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Mefenamic Acid EP Impurity B
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(2,3-Dimethylphenyl)-2-[(2,3-dimethylphenyl)amino]benzamide
CAS 21122-68-9
Analyt- eller komponentnavne N-(2,3-Dimethylphenyl)-2-[(2,3-dimethylphenyl)amino]benzamide
Forsendelsestilstand Room Temperature
Molekylær formel C23 H24 N2 O
Certificeringer/compliance ISO 17025
Synonym Mefenamic Acid Imp. B (EP), N-(2,3-Dimethylphenyl)-2-[(2,3-dimethylphenyl)amino]benzamide, Mefenamic Acid Impurity B
InChI formel InChI=1S/C23H24N2O/c1-15-9-7-13-20(17(15)3)24-22-12-6-5-11-19(22)23(26)25-21-14-8-10-16(2)18(21)4/h5-14,24H,1-4H3,(H,25,26)
SMIL Cc1cccc(NC(=O)c2ccccc2Nc3cccc(C)c3C)c1C
IUPAC navn 2-(2,3-dimethylanilino)-N-(2,3-dimethylphenyl)benzamide
Molekylvægt (g/mol) 344.45
Mængde 100 mg
Formel vægt 344.1889 g/mol