missing translation for 'onlineSavingsMsg'
Få mere at vide

Mefenamic Acid-d3 (major), TRC

Artikelnummer. 30478930
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30478930 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30478930 Supplier Toronto Research Chemicals Supplier No. TRCM2050522.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Mefenamic Acid-d3
CAS 1189707-81-0
Renhedsgrad noter HPLC
Molekylær formel C15 H12 D3 N O2
Mængde 2.5 mg
Synonym 2-[(2,3-Dimethylphenyl)amino]-benzoic Acid-d3, Mefacit-d3, Mefenacid-d3, Mephenamic Acid-d3
InChI formel InChI=1S/C15H15NO2/c1-10-6-5-9-13(11(10)2)16-14-8-4-3-7-12(14)15(17)18/h3-9,16H,1-2H3,(H,17,18)/i5D,6D,9D
SMIL [2H]c1c([2H])c(C)c(C)c(Nc2ccccc2C(=O)O)c1[2H]
IUPAC navn 2-(2,3,4-trideuterio-5,6-dimethyl-anilino)benzoic acid
Molekylvægt (g/mol) 244.30359
Formel vægt 244.1291
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.