Få mere at vide
Mefenacet, TRC
Artikelnummer. 30473639
Skift visning
Mængde:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473639 5 g
30479147 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30473639 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2056005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Mefenacet
CAS 73250-68-7
Molekylær formel C16 H14 N2 O2 S
Mængde 5 g
Synonym 2-(2-Benzothiazolyloxy)-N-methyl-N-phenylacetamide, BTMPA, FOE 1976, Mefenacet, NTN 801, Rancho
InChI formel InChI=1S/C16H14N2O2S/c1-18(12-7-3-2-4-8-12)15(19)11-20-16-17-13-9-5-6-10-14(13)21-16/h2-10H,11H2,1H3
SMIL CN(C(=O)COc1nc2ccccc2s1)c3ccccc3
IUPAC navn 2-(1,3-benzothiazol-2-yloxy)-N-methyl-N-phenylacetamide
Molekylvægt (g/mol) 298.36
Formel vægt 298.0776