missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEF2D Antibody (PCRP-MEF2D-3A4), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14120V
Additional Details : Weight : 0.00970kg
Description
MEF2D Monoclonal antibody specifically detects MEF2D in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
MEF2D | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
4209 | |
Store at 4°C in the dark. | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PCRP-MEF2D-3A4 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
DKFZp686I1536, MADS box transcription enhancer factor 2, polypeptide D (myocyte enhancerfactor 2D), MEF2D/DAZAP1 fusion, myocyte enhancer factor 2D, myocyte enhancer factor 2D/deleted in azoospermia associated protein 1 fusionprotein, myocyte-specific enhancer factor 2D | |
Recombinant full-length human MEF2D protein (Uniprot: Q14814) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG2b |