Få mere at vide
Medroxyprogesterone, Mikromol™
Artikelnummer. 17940606
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
17940606 100 mg
1 options
Artikelnummer. 17940606 Leverandør Mikromol Leverandørnr. MM0635.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Medroxyprogesterone
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Medroxyprogesterone
CAS 520-85-4
Analyt- eller komponentnavne Medroxyprogesterone
Forsendelsestilstand Room Temperature
Molekylær formel C22 H32 O3
Certificeringer/compliance ISO 17034
Synonym Medroxyprogesterone Acetate Imp. B (EP), Medroxyprogesterone Acetate USP Related Compound B, Medroxyprogesterone Acetate USP RC B, Medroxyprogesterone, 17-Hydroxy-6alpha-methylpregn-4-ene-3,20-dione, Pregn-4-ene-3,20-dione, 17-hydroxy-6-methyl-, (6alpha)-, Pregn-4-ene-3,20-dione, 17-hydroxy-6alpha-methyl- (7CI,8CI), Progesterone, 17-hydroxy-6alpha-methyl- (6CI), (6alpha)-17-Hydroxy-6-methylpregn-4-ene-3,20-dione, 17-Hydroxy-6alpha-methylprogesterone, 17alpha-Hydroxy-6alpha-methylprogesterone, 6alpha-Methyl-17-hydroxyprogesterone, 6alpha-Methyl-17R-hydroxyprogesterone, 6alpha-Methyl-17alpha-hydroxypregn-4-ene-3,20-dione, 6alpha-Methyl-17alpha-hydroxyprogesterone, 6alpha-Methyl-4-pregnen-17alpha-ol-3,20-dione, Medoxyprogesterone, Medroxyprogesteron, NSC 27408, U 8840, Medroxyprogesterone Acetate Related Compound B, Medroxyprogesterone Acetate Impurity B
InChI formel InChI=1S/C22H32O3/c1-13-11-16-17(20(3)8-5-15(24)12-19(13)20)6-9-21(4)18(16)7-10-22(21,25)14(2)23/h12-13,16-18,25H,5-11H2,1-4H3/t13-,16+,17-,18-,20+,21-,22-/m0/s1
SMIL C[C@H]1C[C@@H]2[C@H](CC[C@@]3(C)[C@H]2CC[C@]3(O)C(=O)C)[C@@]4(C)CCC(=O)C=C14
IUPAC navn (6S,8R,9S,10R,13S,14S,17R)-17-acetyl-17-hydroxy-6,10,13-trimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 344.49
Mængde 100 mg
Formel vægt 344.2351 g/mol