Få mere at vide
Medroxy Progesterone-d6 17-Acetate, TRC
Artikelnummer. 30472193
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30472193 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472193 Leverandør Toronto Research Chemicals Leverandørnr. TRCM20356210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Medroxyprogesterone 17-acetate-d6 (6alpha-methyl-d3,acetyl-d3)
CAS 2376036-01-8
Renhedsgrad noter HPLC
Molekylær formel C24 2H6 H28 O4
Mængde 10 mg
Synonym Pregn-4-ene-3,20-dione, 17-(acetyl-2,2,2-d3-oxy)-6-(methyl-d3)-, (6α)-, (6α)-17-(Acetyl-2,2,2-d3-oxy)-6-(methyl-d3)-pregn-4-ene-3,20-dione, Medroxy progesterone-d6 17-acetate, Medroxyprogesterone-d6 acetate, Medroxyprogesterone Acetate D6 (6α-methyl-D3,acetyl-D3)
InChI formel InChI=1S/C24H34O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h13-14,18-20H,6-12H2,1-5H3/t14-,18+,19-,20-,22+,23-,24-/m0/s1/i1D3,3D3
SMIL [2H]C([2H])([2H])[C@H]1C[C@@H]2[C@H](CC[C@@]3(C)[C@H]2CC[C@]3(OC(=O)C([2H])([2H])[2H])C(=O)C)[C@@]4(C)CCC(=O)C=C14
IUPAC navn [(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-10,13-dimethyl-3-oxo-6-(trideuteriomethyl)-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] 2,2,2-trideuterioacetate
Molekylvægt (g/mol) 392.56
Formel vægt 392.2834
Procent renhed >95