Få mere at vide
Medroxy Progesterone-d3, TRC
Artikelnummer. 30472639
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30472639 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472639 Leverandør Toronto Research Chemicals Leverandørnr. TRCM20355210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Medroxyprogesterone-d3
CAS 162462-69-3
Renhedsgrad noter HPLC
Molekylær formel C22H29D3O3
Mængde 10 mg
Synonym Medroxyprogesterone D3 (6-Methyl D3), (6α)-17-Hydroxy-6-(methyl-d3)pregn-4-ene-3,20-dione, 6β,7-Dihydro-6-(methyl-d3)megestrol
InChI formel InChI=1S/C22H32O3/c1-13-11-16-17(20(3)8-5-15(24)12-19(13)20)6-9-21(4)18(16)7-10-22(21,25)14(2)23/h12-13,16-18,25H,5-11H2,1-4H3/t13-,16+,17-,18-,20+,21-,22-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])[C@H]1C[C@@H]2[C@H](CC[C@@]3(C)[C@H]2CC[C@]3(O)C(=O)C)[C@@]4(C)CCC(=O)C=C14
Molekylvægt (g/mol) 347.51
Procent renhed >95