Learn More
Corning™ Medium 199 (Mod.)
Often used in vaccine production, virology, and for the culture of various non-transformed cells. It was originally formulated as the result of efforts to create a fully defined, animal origin-free medium.
POA
Description
Product manufactured under industry-leading standards for low endotoxin content and superior results. Medium 199 is often used in vaccine production, virology, and for the culture of various non-transformed cells. It was originally formulated as the result of efforts to create a fully defined, animal origin-free medium. Long-term use of this medium requires serum supplementation. This formulation contains Earle's salts and L-glutamine, and it does not contain sodium bicarbonate.Specifications
2° to 8°C | |
2° to 8°C | |
10000 mL |