Learn More
Gibco™ Human Fibroblast Expansion Basal Medium (formerly Medium 106)
GREENER_CHOICE
Human Fibroblast Expansion Basal Medium (formerly Medium 106) is a sterile, liquid tissue culture medium intended for use as one component in a complete culture environment for the growth of normal human dermal fibroblasts.
Brand:  Gibco™ M106500
Product Code. 11607094
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
This basal culture medium contains:
  • Essential and non-essential amino acids
  • Vitamins
  • Other organic compounds
  • Trace minerals
  • Inorganic salts.
Human Fibroblast Expansion Basal Medium does NOT contain:
  • Antibiotics
  • Antimycotics
  • Hormones
  • Growth factors
  • Proteins
This medium is HEPES and bicarbonate buffered and is designed for use in an incubator with an atmosphere of 5% CO2 / 95% air. To support the plating and long-term proliferation of human dermal fibroblasts, Human Fibroblast Expansion Basal Medium must be supplemented with Low Serum Growth Supplement (LSGS, Cat. No. S-003-10), or Low Serum Growth Supplement Kit (LSGS Kit, Cat. No. S-003-K).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fibroblast Cells
Medium 106
Sterile-filtered
Cascade Biologics™
Room Temperature
Mammalian Cell Culture
Liquid
Sterile-filtered
HEPES, Phenol Red
500 mL
Store at 4°C.
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Gibco™ Human Fibroblast Expansion Basal Medium (formerly Medium 106) > Quantity: 500 mL