Få mere at vide
Corning™ Media Tech™ Yeast Extract Powder
BIOPHARMA
Artikelnummer. 15363761
Skift visning
Mængde:
500 g
Pakningsstørrelse:
500g
Artikelnummer. Mængde unitSize
15363761 500 g 500g
1 options
Artikelnummer. 15363761 Leverandør Corning™ Leverandørnr. 61231RO
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Corning classical growth media products are complemented with a comprehensive range of exceptionally high quality nutrients, supplements, buffers, antiobiotics, antimycotics and other reagents specially formulated to maximise results and consistency in your cell culture processes.
TRUSTED_SUSTAINABILITY
Tekniske data
Produkttype Yeast
Mængde 500 g