Få mere at vide
AMPri™ Med-Comfort Premium Grip Latex Examination Gloves, PPE Cat. III
Beskrivelse
- AQL 1.5
- Premium grip examination gloves
- Applications include laboratory, clinical, pharmaceutical, dental, and food industry use
- Protect against chemicals and microorganisms
- Ambidextrous fit
- Powder-free
- Non-sterile
- Color: white
- Textured finish
- Disposable
Tekniske data
Tekniske data
| Certificeringer/compliance | AQL 1.5 |
| Certificeringer/overholdelse (Europa) | EN 455-1, EN374-1/4/5, EN420, Food grade |
| Farve | White |
| Engangs | Single-use |
| Brug af laboratorium/renrum | Laboratory |
| Latex fri | No |
| Materiale | Latex |
| Størrelse | Large |
| Tykkelse (metrisk) | 0.08 mm (Cuff) |
| Længde (metrisk) | 240 mm |