Få mere at vide
Meclofenamic Acid-d4, TRC
Artikelnummer. 30472641
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30472641 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472641 Leverandør Toronto Research Chemicals Leverandørnr. TRCM20275210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Meclofenamic Acid-d4
CAS 1185072-18-7
Molekylær formel C14H7D4Cl2NO2
Mængde 10 mg
InChI formel InChI=1S/C14H11Cl2NO2/c1-8-6-7-10(15)13(12(8)16)17-11-5-3-2-4-9(11)14(18)19/h2-7,17H,1H3,(H,18,19)/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(C(=O)O)c(Nc2c(Cl)ccc(C)c2Cl)c1[2H]
Molekylvægt (g/mol) 300.17