Få mere at vide
Meclizine N,N’-Dioxide, TRC
Artikelnummer. 30472144
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30472144 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472144 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2027055MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Meclizine N,N'-Dioxide
CAS 114624-70-3
Molekylær formel C25 H27 Cl N2 O2
Mængde 5 mg
Synonym 1-[(4-Chlorophenyl)phenylmethyl]-4-[(3-methylphenyl)methyl]piperazine 1,4-Dioxide
InChI formel InChI=1S/C25H27ClN2O2/c1-20-6-5-7-21(18-20)19-27(29)14-16-28(30,17-15-27)25(22-8-3-2-4-9-22)23-10-12-24(26)13-11-23/h2-13,18,25H,14-17,19H2,1H3
SMIL Cc1cccc(C[N+]2([O-])CC[N+]([O-])(CC2)C(c3ccccc3)c4ccc(Cl)cc4)c1
IUPAC navn 1-[(4-chlorophenyl)-phenylmethyl]-4-[(3-methylphenyl)methyl]-1,4-dioxidopiperazine-1,4-diium
Molekylvægt (g/mol) 422.95
Formel vægt 422.1761