Få mere at vide
Meclizine Dihydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Meclozine Hydrochloride |
| CAS | 1104-22-9 |
| Molekylær formel | C25 H27 Cl N2 . 2 Cl H |
| Mængde | 1 g |
| Synonym | Meclozine Dihydrochloride, 1-[(4-Chlorophenyl)phenylmethyl]-4-[(3-methylphenyl)methyl]piperazine dihydrochloride, 1-(p-Chloro-α-phenylbenzyl)-4-(m-methylbenzyl)piperazine dihydrochloride, Ancolan dihydrochloride, Bonamine, Diadril, Meclizine dihydrochloride, Meclizine hydrochloride, Meclozine dihydrochloride, Meclozine hydrochloride, NSC 28728, Navidoxine, Parachloramine dihydrochloride, Postafen, Postafene dihydrochloride |
| InChI formel | InChI=1S/C25H27ClN2.2ClH/c1-20-6-5-7-21(18-20)19-27-14-16-28(17-15-27)25(22-8-3-2-4-9-22)23-10-12-24(26)13-11-23;;/h2-13,18,25H,14-17,19H2,1H3;2*1H |
| SMIL | Cl.Cl.Cc1cccc(CN2CCN(CC2)C(c3ccccc3)c4ccc(Cl)cc4)c1 |
| IUPAC navn | 1-[(4-chlorophenyl)-phenylmethyl]-4-[(3-methylphenyl)methyl]piperazine;dihydrochloride |
| Molekylvægt (g/mol) | 463.87 |
| Formel vægt | 462.1396 |