Få mere at vide
Meclizine-d8 Dihydrochloride, TRC
Artikelnummer. 30477322
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30477322 10 mg
30472194 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30477322 Leverandør Toronto Research Chemicals Leverandørnr. TRCM20270210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1246816-06-7
Molekylær formel C25H21D8Cl3N2
Molekylvægt (g/mol) 471.92
Synonym 1-[(4-Chlorophenyl)phenylmethyl]-4-[(3-methylphenyl)methyl](piperazine-d8) DiHydrochloride, UCB-5062-d8, Ancolan-d8, Antivert-d8, Bonamine-d8, Bonine-d8, Calmonal-d8, Permesin-d8, Postafen-d8
SMIL CC1=CC(CN(C([2H])([2H])C2([2H])[2H])C([2H])([2H])C([2H])([2H])N2C(C3=CC=CC=C3)C4=CC=C(Cl)C=C4)=CC=C1
Tekniske data
Kemisk navn eller materiale Meclizine-d8 Dihydrochloride
Mængde 10 mg
InChI formel InChI=1S/C25H27ClN2/c1-20-6-5-7-21(18-20)19-27-14-16-28(17-15-27)25(22-8-3-2-4-9-22)23-10-12-24(26)13-11-23/h2-13,18,25H,14-17,19H2,1H3/i14D2,15D2,16D2,17D2