Få mere at vide
Mecamylamine-d3 Hydrochloride, TRC
Artikelnummer. 30467003
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30467003 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467003 Leverandør Toronto Research Chemicals Leverandørnr. TRCM20260210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Mecamylamine hydrochloride N-methyl-d3
CAS 1217625-71-2
Molekylær formel C11 2H3 H18 N . Cl H
Mængde 10 mg
Synonym 2,3,3-Trimethyl-N-(trideuteriomethyl)norbornan-2-amine hydrochloride, Mecamylamine-D3 hydrochloride (N-methyl D3)
InChI formel InChI=1S/C11H21N.ClH/c1-10(2)8-5-6-9(7-8)11(10,3)12-4;/h8-9,12H,5-7H2,1-4H3;1H/i4D3;
SMIL Cl.[2H]C([2H])([2H])NC1(C)C2CCC(C2)C1(C)C
IUPAC navn 2,2,3-trimethyl-N-(trideuteriomethyl)bicyclo[2.2.1]heptan-3-amine;hydrochloride
Molekylvægt (g/mol) 206.77
Formel vægt 206.1629