missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Mebhydrolin napadisylate, MedChemExpress
Description
Mebhydrolin napadisylate is a specific histamine H1 receptor antagonist.
Specifications
Specifications
| Kemisk navn eller materiale | Mebhydrolin napadisylate |
| CAS | 6153-33-9 |
| Farve | White |
| Renhedsgrad noter | Research |
| Molekylær formel | C48H48N4O6S2 |
| Mængde | 100 mg |
| Synonym | Mebhydroline 1,5-naphthalenedisulfonate salt |
| Opløselighedsinformation | DMSO : 5.56 mg/mL (6.61 mM; ultrasonic and warming and heat to 60°C) |
| SMIL | O=S(C1=C2C=CC=C(S(=O)(O)=O)C2=CC=C1)(O)=O.CN(C3)CCC(N4CC5=CC=CC=C5)=C3C6=C4C=CC=C6.CN(C7)CCC(N8CC9=CC=CC=C9)=C7C%10=C8C=CC=C%10 |
| Molekylvægt (g/mol) | 841.05 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?