Få mere at vide
Mebendazole-d8, TRC
Artikelnummer. 30469286
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30469286 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469286 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2005021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Mebendazole D3 (methyl D3)
CAS 1173021-87-8
Molekylær formel C16 2H3 H10 N3 O3
Mængde 1 mg
Synonym Trideuteriomethyl N-(5-benzoyl-1H-benzimidazol-2-yl)carbamate, Mebendazole D3, Mebendazole D3 (methyl D3)
InChI formel InChI=1S/C16H13N3O3/c1-22-16(21)19-15-17-12-8-7-11(9-13(12)18-15)14(20)10-5-3-2-4-6-10/h2-9H,1H3,(H2,17,18,19,21)/i1D3
SMIL [2H]C([2H])([2H])OC(=O)Nc1nc2cc(ccc2[nH]1)C(=O)c3ccccc3
IUPAC navn trideuteriomethyl N-(5-benzoyl-1H-benzimidazol-2-yl)carbamate
Molekylvægt (g/mol) 298.31
Formel vægt 298.1145