Learn More
WTW™ Measuring Bottle
Product Code. 10243391
Change view
Click to view available options
Volume (Metric):
250 mL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
10243391 250 mL Each
1 options
Product Code. 10243391 Supplier WTW™ Supplier No. 209106
Forventet forsendelse: 11-02-2027
WTW offers a wide variety of measuring bottles and measuring vessels with adapters to measure BSB, soil respiration, degradeability as per OECD 301F, fermentation tests, biogas and much more. They have all be tested in their respective applications and therefore support accurate measurements.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 250 mL
For Use With (Equipment) Respirometry