Få mere at vide
TMC Hallcrest™ Thermax Chillchecker Temperature Indicators
Vise:
-17 °C
-2 °C
-8 °C
9 °C
Unit Size:
Pakke med 100
Description
- Confirmation that the temperatures have not been exceeded.
- Color changes to blue when temperature is reached.
- Easy to use and read.
- Self Adhesive.
- Available temperatures between -17 to 20°C.
- Accurate to +/-1°C
Specifications
Specifications
| Dybde (engelsk) | 0.23 in. |
| Dybde (metrisk) | 6 mm |
| Engangs | Single-use |
| Materiale | PVC |
| Bredde (engelsk) | 1.25 in. |
| Produkttype | Temperature Indicator |
| Nøjagtighed | ±1°C |
| Vise | -17 °C |
| Bredde (metrisk) | 32 mm |