Få mere at vide
MDV 3100-d3, TRC
Artikelnummer. 30484216
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30484216 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30484216 Leverandør Toronto Research Chemicals Leverandørnr. TRCM1998021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale MDV 3100-d3
CAS 1443331-82-5
Molekylær formel C21H13D3F4N4O2S
Mængde 1 mg
Synonym 4-[3-[4-Cyano-3-(trifluoromethyl)phenyl]-5,5-dimethyl-4-oxo-2-thioxo-1-imidazolidinyl]-2-fluoro-N-(methyl-d3)benzamide, Enzalutamide-d3
InChI formel InChI=1S/C21H16F4N4O2S/c1-20(2)18(31)28(12-5-4-11(10-26)15(8-12)21(23,24)25)19(32)29(20)13-6-7-14(16(22)9-13)17(30)27-3/h4-9H,1-3H3,(H,27,30)/i3D3
SMIL FC1=C(C(NC([2H])([2H])[2H])=O)C=CC(N(C(N2C3=CC=C(C#N)C(C(F)(F)F)=C3)=S)C(C)(C)C2=O)=C1
Molekylvægt (g/mol) 467.45