Få mere at vide
Novus Biologicals™ MDH2 Overexpression Lysate
Artikelnummer. 18250545 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mg
Artikelnummer. Mængde
18250545 0.1 mg
1 options
Artikelnummer. 18250545 Leverandør Novus Biologicals™ Leverandørnr. NBL112966
 Begrænset vare
Forventet forsendelse: 09-10-2026
The MDH2 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Tekniske data
Cellelinje The lysate was created in HEK293T cells, using plasmid ID RC2195 and based on accession number NM_5918. The protein contains a C-terminal DDK tag.
Til brug med (applikation) Western Blot
Mængde 0.1 mg
Opbevaringsbuffer RIPA buffer
Renhed Protein
Testspecificitet Homo sapiens malate dehydrogenase 2, NAD (mitochondrial) (MDH2), nuclear gene encoding mitochondrial protein, mRNA.
Indhold og opbevaring -80°C. Avoid freeze-thaw cycles.
Forskningskategori Signal Transduction
Proteintilstand Native
For Research Use Only